Root Tank
Actual product may vary
Fisherbrand
Root Tank
Light: No Lighting
Have Questions?
Catalog NumberColor
FISS06679Clear
Catalog number FISS06679
Price (SGD)
-
Color:
Clear
When plants are placed in the root tank, the roots spread out to seize as much water as possible. When they encounter the sloped wall, they spread along it, providing a clear and dramatic view of the intricate root systems. Integrated tray catches run-off from watering, limiting the mess.
  • Welded acrylic durable construction
  • Slides slope at 20° angle
  • Draining holes at bottom
  • Can be used with Greenbox Gardening System (S06623)
  • Dimensions: 10 x 4.5 x 8 in. (25.4 x 11.4 x 20.3cm)
Specifications
ColorClear
DescriptionRoot Tank
LightingNo Lighting
MaterialAcrylic
TypeRoot Tank
Dimensions (L x W x H)10 x 4.5 x 8 in.
Unit SizeEach