Porosity of Soils
Porosity of Soils
Actual product may vary
Fisher Catalogue
Porosity of Soils
Kit shows the relationship between particle size, shape and uniformity.
Have Questions?
Catalog number FISS05974
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Place two different soil samples in the side-by-side high-quality tubes
  • Using the included syringes, insert an equal amount of water in each
  • Measure the rate of flow to determine porosity
  • Fully assembled unit has precision-cut sytrene housing
Specifications
DescriptionPorosity of Soils Apparatus
IncludesHigh-quality tubes, syringes, precision-cut sytrene housing
Unit SizeEA