Check Order Status
Rubber Bands for Science Kits
Actual product may vary
Rubber Bands for Science Kits
Consumables used with multiple projects and science kits.
Have Questions?
Change view
Catalog Number
Size
FISS05370
33
FISS05212
16
2 Options
Catalog number
FISS05370
Price (SGD)
-
Size:
33
Specifications
Product Type
Rubber Band
Material
Rubber
For Use With (Application)
For multiple projects or in a science kit
Size
33
Weight (English)
0.25 lb.
Weight (Metric)
0.113 kg