Balloons for Science Kits
Actual product may vary
Fisherbrand
Balloons for Science Kits
Consumbles for use with multiple projects and science kits.
Have Questions?
Catalog NumberColorDiameter (English) OuterShape
FISS05357Dark Blue9 in.Round
Catalog number FISS05357
Price (SGD)
-
Color:
Dark Blue
Diameter (English) Outer:
9 in.
Shape:
Round
  • Round
  • Oblong
Specifications
Product TypeBalloon
ColorDark Blue
For Use With (Application)For use in multiple projects and in science kits
ShapeRound
Diameter (English) Outer9 in.
Diameter (Metric) Outer23 cm
Unit SizePack