Crucibles
Crucibles
Actual product may vary
Crucibles
Crucibles
Crucibles
Easy to clean smooth, glazed surface.
Have Questions?
Catalog NumberCapacity (Metric)Product Type
FISS0222230 mLHigh-form
Catalog number FISS02222
Price (SGD)
-
Capacity (Metric):
30 mL
Product Type:
High-form
  • Use in any chemical environment
  • Withstand temperatures up to 1150°C (2,102°F)
  • Consistent wall thickness and density
  • Fired twice to give added strength and durability
  • Highly resistant to chemical attack
Specifications
MaterialPorcelain
FinishSmooth, Glazed
For Use With (Application)Used in chemical environments
Max. Temperature (Metric)1150°C
Capacity (Metric)30 mL
Product TypeHigh-form
Unit SizePK