Starch Solution R, EP Reagent 1085103, Ricca Chemical
Starch Solution R, EP Reagent 1085103, Ricca Chemical
Starch Solution R, EP Reagent 1085103, Ricca Chemical
Starch Solution R, EP Reagent 1085103, Ricca Chemical
Starch Solution R, EP Reagent 1085103, Ricca Chemical
Have Questions?
Catalog NumberQuantity
FISR8046000500500 mL
Catalog number FISR8046000500
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5, 9005-84-9, 7774-29-0
Specifications
Chemical Name or MaterialStarch Solution
ColorUndesignated
Name NoteReagent
CAS7732-18-5
CAS Min %97.0
View more