Check Order Status
Potassium Chloride Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Element Search
Structure Search
Potassium Chloride Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FISR5888141500
500 mL
Catalog number
FISR5888141500
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Chloride Conductivity Standard
Shelf Life
18 months
Name Note
1, 413 μmho/cm at 25°C
Packaging
Natural Poly Bottle
CAS
7447-40-7
View more