Check Order Status
Phosphorus Standard, 10 ppb (10 μg/L) P, Ricca Chemical
Element Search
Structure Search
Phosphorus Standard, 10 ppb (10 μg/L) P, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FISR58561001A
1 L
Catalog number
FISR58561001A
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material
Phosphorus Standard
Shelf Life
6 months
Packaging
Natural Poly Bottle
CAS
7778-77-0
CAS Min %
98.0
View more