Check Order Status
Copper Tetrammine, Ammoniacal Solution, Ricca Chemical
Element Search
Structure Search
Copper Tetrammine, Ammoniacal Solution, Ricca Chemical
EP Reagent 1022600
Have Questions?
Catalog Number
Quantity
FISR2370000250
250 mL
Catalog number
FISR2370000250
Price (SGD)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Copper Tetrammine
Color
Blue
Name Note
Ammoniacal Solution
CAS
1336-21-6
CAS Min %
54.0
View more