Check Order Status
Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISR161016
500 mL
Clear PET-G Bottle
Catalog number
FISR161016
Price (SGD)
-
Quantity:
500 mL
Packaging:
Clear PET-G Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7558-79-4, 1310-73-2, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Shelf Life
18 months
Name Note
pH 11.00± 0.01 at 25°C
CAS
7558-79-4
Melting Point
0°C
View more