Check Order Status
Petri Dishes and Petri-Pad™ Dishes
Actual product may vary
MilliporeSigma
Petri Dishes and Petri-Pad™ Dishes
Available with or without absorbent pads
Have Questions?
Change view
Catalog Number
Type
Quantity
FISPD20047S0
Dish with Pad
150 Pack
FISPD2004700
Dish without Pad
100 Pk. (5 Sleeves)
2 Options
Catalog number
FISPD20047S0
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Type:
Dish with Pad
Quantity:
150 Pack
Available with or without absorbent pads
Petri Dishes
47mm sterilized Petri dishes
Use with solid agar medium
Dishes are made of molded polystyrene
Petri-Pad Dishes
Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
Eliminate labor and cost of hand-loading pads and minimize contamination
Pads are pure cellulose
Specifications
Color
Clear
Disposable
Single-use
Material
Polystyrene
Shape
Round
Sterility
Sterile
Type
Dish with Pad
Packaging
Sleeve
Quantity
150 Pack
Diameter (Metric)
47 mm