Owl™ Dual-Gel Vertical Electrophoresis Systems
Actual product may vary
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Thermo Scientific
Owl™ Dual-Gel Vertical Electrophoresis Systems
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
Have Questions?
Catalog NumberVolume (Metric) BufferIncludes
FISOW-P10DS11250 mLComplete System plus Caster
Catalog number FISOW-P10DS1
Price (SGD)
-
Volume (Metric) Buffer:
1250 mL
Includes:
Complete System plus Caster
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel
Features:
  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
Specifications
Product LineOwl
TypeDual-Gel Vertical Electrophoresis Complete System with Caster
Gel Dimensions20 x 20  cm
IncludesComplete System plus Caster
ModelP10DS
Volume (Metric) Buffer1250 mL
Length (English)12 in.
Length (Metric) Gel20 cm
Width (English)6.5 in.
Width (Metric) Gel20 cm
Height (English)9 in.
Unit SizeEach