Omnifit™ T-Series Bottle Caps
Omnifit™ T-Series Bottle Caps
Actual product may vary
Kinesis
Omnifit™ T-Series Bottle Caps
Easy to use and versatile range of caps requires no fittings
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThread Size (Metric)Includes
FISOF945T3FGL 453 Ports, Check Valve
FISOF945T2GL 452 Ports, No Valve
Catalog number FISOF945T3F
Price (SGD)
288.00
Each
Add to cart
Thread Size (Metric):
GL 45
Includes:
3 Ports, Check Valve
Price (SGD)
288.00
Each
Add to cart
Easy to use and versatile range of caps requires no fittings
  • Luer ported, straight through tubing
  • Simply push 1/8 in. O.D. hard wall tubing through the ports
  • For 1/16 in. O.D. hard wall tubing, use OF9LA adapter supplied with cap
  • Use OF9BA series adapter for softwall tubing
  • Anti-twist design prevents tubing from kinking when removing cap
  • Each port also accepts male Luer connectors
  • With polypropylene screw cap and chemically inert solid PTFE body
Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.
Specifications
ColorBlue, White
Includes3 Ports, Check Valve
MaterialPTFE
No. per Case1 Cs.
SterilityNon-sterile
Thread Size (Metric)GL 45
DisposableReusable
Height (Metric)29.8 mm
Product TypeSolvent Waste Cap
For Use With (Equipment)T series solvent bottle
Diameter (Metric)54 mm