Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Histology and Cytology
▸
Slide Mounting Media
▸
Fluoromount-G™ Slide Mounting Medium
Actual product may vary
SouthernBiotech
Fluoromount-G™ Slide Mounting Medium
Water soluble, nonfluorescing mounting medium
Have Questions?
Catalog Number
Volume (Metric)
FISOB100-01
25 mL
Catalog number
FISOB100-01
Price (SGD)
-
Volume (Metric):
25 mL
Ideal for immunofluorescent staining procedures
Particularly well-suited for slide staining procedures with an aqueous final step
Supplied in sufficient quantity to mount approximately 500 slides
Specifications
Applications
Immunofluorescent Staining Procedures
Volume (English)
0.84 oz.
Volume (Metric)
25 mL