Check Order Status
Mediatech™ Cell Culture Phosphate Buffered Saline (10X)
Actual product may vary
Mediatech™ Cell Culture Phosphate Buffered Saline (10X)
With low endotoxin content
Have Questions?
Catalog Number
Quantity
FISMT-46013CM
6 x 1000 mL
Catalog number
FISMT-46013CM
Price (SGD)
1,017.00
CS
Add to cart
Quantity:
6 x 1000 mL
Price (SGD)
1,017.00
CS
Add to cart
Maintains cell culture media in the physiological pH range
Maintains cell tonicity and viability
Produced under the highest industry standards to ensure superior results
Nontoxic
Liquid without calcium and magnesium
Specifications
Additive
No Calcium, No Magnesium
Dilution
10 X
Quantity
6 x 1000 mL
Formulation
Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2.40g/L, Sodium Chloride 80g/L, Disodium Phosphate 14.4g/L
pH
7.3 to 7.5
Form
Liquid
Unit Size
CS
Contents & Storage
15° to 30°C