Check Order Status
Ammonium Hydroxide ACS, MilliporeSigma™
Element Search
Structure Search
Ammonium Hydroxide ACS, MilliporeSigma™
Have Questions?
Catalog Number
Quantity
Packaging
FISMAX13036
500 mL
Glass Bottle
Catalog number
FISMAX13036
Price (SGD)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ammonium Hydroxide
Name Note
EMSURE™, For Analysis
CAS
1336-21-6
Recommended Storage
Store below +25°C
Assay Percent Range
28 to 30%
View more