Check Order Status
Manganese Sulfate Monohydrate (Powder/Technical), Fisher Chemical
Element Search
Structure Search
Manganese Sulfate Monohydrate (Powder/Technical), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISM118-3
3 kg
Poly Bottle
Catalog number
FISM118-3
Price (SGD)
-
Quantity:
3 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Manganese Sulfate
CAS
10034-96-5
ChemAlert Storage Symbol
Gray
Physical Form
Solid
Quantity
3 kg
View more