1x1 image pixel for data collection
Precoated TLC and PLC Glass Plates
Precoated TLC and PLC Glass Plates
Actual product may vary
MilliporeSigma
Precoated TLC and PLC Glass Plates
Average particle size is 15μm with a 5-20μm distribution
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)QuantityMaterialType
FISM116485000120 x 20 cm25/Pk.Silica Gel 60 W F254 sClassical Silica TLC Plate
FISM105721000120 x 20 cm25/Pk.Silica 60Unmodified Silica Gel TLC Plate
Catalog number FISM1164850001
Price (SGD)
1,464.00
EA
Add to cart
Dimensions (L x W):
20 x 20 cm
Quantity:
25/Pk.
Material:
Silica Gel 60 W F254 s
Type:
Classical Silica TLC Plate
Price (SGD)
1,464.00
EA
Add to cart
  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates
Specifications
Analysis Time20 to 200 min.
Backing MaterialGlass
Certifications/ComplianceISO 9001:2000
MaterialSilica Gel 60 W F254 s
Particle Size10 to 12 μm
Pore Size60 Å
Quantity25/Pk.
TypeClassical Silica TLC Plate
Excitation Wavelength254 nm (Acid Stable)
For Use With (Application)Versatile and Reliable Routine Analysis of a Broad Range of Substances
No. of Samples10.0
Particle Size Distribution5 to 20 μm
Pore Volume0.8 mL/g
SorbentSilica Gel 60 W F254 s
Specific Surface520 m2/g
Thickness (Metric)250 μm
Dimensions (L x W)20 x 20 cm
Length (Metric)20 cm
Width (Metric)20 cm
Height (English)0.0011 in.
Height (Metric)30 μm
Unit SizeEA
Share catalog number, name or link