TLC LuxPlate™ Silica Gel 60 F254: 25 Glass plates, 20 x 20cm
TLC LuxPlate&trade; Silica Gel 60 F<sub>254</sub>: 25 Glass plates, 20 x 20cm
Actual product may vary
MilliporeSigma
TLC LuxPlate™ Silica Gel 60 F254: 25 Glass plates, 20 x 20cm
Allows analyzation of any substance when combined with suitable mobile phase
Have Questions?
Catalog NumberDimensions (L x W)
FISM105805000120 x 20 cm
Catalog number FISM1058050001
Price (SGD)
981.00
EA
Add to cart
Dimensions (L x W):
20 x 20 cm
Price (SGD)
981.00
EA
Add to cart
  • Combination of unmodified silica gel 60 and polymeric binder for hard and adherent surface, resistant to crack and damage
  • Dense plate coating ensuring narrow bands and maximum sample separation efficiency
  • Layer thickness of 250μm with mean particle size of 10 to 12μm
  • Green fluorescencing inorganic florescent indications for UV detection of colorless substance
  • Disposable
  • Fibre case packaging
  • In-process control in drug synthesis
  • Identity and stability testing of drugs
  • Quality control of pharmaceuticals, food and environmental samples
  • Residue analysis in food and environmental samples
Specifications
Analysis Time20 to 200 min.
Backing MaterialGlass
Particle Size10 to 12 μm
Pore Size60 Å
Quantity25/Pk.
TypeClassical Silica TLC Plate
Excitation Wavelength254 nm
For Use With (Application)In-Process Control in Drug Synthesis, Identity and Stability Testing of Drugs, Quality Control of Pharmaceuticals, Food and Environmental Samples, Residue Analysis in Food and Environmental Samples
No. of Samples10.0
Particle Size Distribution5 to 20 μm
Pore Volume0.8 mL/g
SorbentSilica Gel 60 F254
Specific Surface520 m2/g
Thickness (Metric)250 μm
Dimensions (L x W)20 x 20 cm
Length (Metric)20 cm
Width (Metric)20 cm
Height (English)0.0011 in.
Height (Metric)30 μm
Unit SizeEA