Mitex™ Membrane Filters, 5 μm
Mitex™ Membrane Filters, 5 μm
Actual product may vary
MilliporeSigma
Mitex™ Membrane Filters, 5 μm
Unlaminated for extreme chemical or temperature conditions
Have Questions?
Catalog NumberDiameter (Metric)
FISLSWP14250142 mm
Catalog number FISLSWP14250
Price (SGD)
3,506.00
PK
Add to cart
Diameter (Metric):
142 mm
Price (SGD)
3,506.00
PK
Add to cart
Unlaminated for extreme chemical or temperature conditions
Mitex™ PTFE Membrane is unaffected by almost all liquids, including organic solvents, concentrated acids and bases, propellants, and cryogenic fluids. Mitex™ membrane is unbacked, yet is easy to handle, combining the convenience of a backed membrane with the versatility of a pure PTFE membrane. Mitex™ membrane is hydrophobic.
Features & Benefits:
  • Broad chemical compatibility
  • Biologically and chemically inert
  • Stable at temperatures in excess of 260 °C (500 °F) and below -100 °C (-148 °F)*
Applications: Clarifying Acids, Bases, and Cryogenic Fluids; Clarifying Propellants; Analyzing Hydraulic Fluids; Isolating RNA
Specifications
AutoclavableNot Autoclavable
Bubble Point0.9 psi
ColorWhite
Filter TypeScreen Filter
MaterialPTFE
Pore Size5 μm
SterilityNon-sterile
Flow Rate70 mL/min x cm2
For Use With (Application)Clarification of acids, bases, and cryogenic fluids; clarification of propellants; analysis of hydraulic fluids; RNA isolation.
Membrane TypePlain
No. per Case50Pack
Quantity50/Pk.
Thickness (Metric)125 μm
TypeFilter Membranes
WettabilityHydrophobic
Diameter (Metric)142 mm
Unit SizePK