SoluLyse™ Bacterial Protein Extraction Reagent
Actual product may vary
Genlantis
SoluLyse™ Bacterial Protein Extraction Reagent
Efficient and gentle extraction of soluble proteins from bacteria
Have Questions?
Catalog NumberQuantityFormulation
FISL200500500 mL20mM Tris HCl Buffer, pH 7.5
Catalog number FISL200500
Price (SGD)
-
Quantity:
500 mL
Formulation:
20mM Tris HCl Buffer, pH 7.5
  • Produces high yield of soluble proteins
  • Simple one-step protocol
  • Proprietary formulation of nonionic detergents optimized for bacterial protein extraction
  • Proteins retain more activity
  • Compatible with protease inhibitors, salts, chelating agents and reducing agents
  • Choice of Phosphate or Tris buffer formulations to avoid interference with affinity purification resins and other downstream applications
Specifications
Formulation20mM Tris HCl Buffer, pH 7.5
Quantity500 mL
Product TypeBacterial Protein Extraction Reagent
For Use With (Application)Proprietary formulation of nonionic detergents optimized for bacterial Protein extraction, high Protein yield with simple one-step proto col
Unit SizeEach
Contents & Storage
Room Temperature