Check Order Status
Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Filtration/Delivery Four-Valve Cap System
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Filtration/Delivery Four-Valve Cap System
Parts and accessories for standard four-valve cap system
Have Questions?
Catalog Number
Product Type
FISK953981-0004
Retainer Ring
Catalog number
FISK953981-0004
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Retainer Ring
Lower cap retainer ring
Valve stem, CTFE, short (for sparge and pump ports)
Check valve with retainer
Lower screw cap, GL 45
Specifications
No. per Case
1 Cs.
Product Type
Retainer Ring
For Use With (Equipment)
Ultra-ware™ standard filtration/delivery four valve cap system