1x1 image pixel for data collection
Kimble™ Kimble™ Kontes™ Components for Ultra-Ware™ Filtration Cap System
Kimble™ Kimble™ Kontes™ Components for Ultra-Ware™ Filtration Cap System
Actual product may vary
DWK Life Sciences
Kimble™ Kimble™ Kontes™ Components for Ultra-Ware™ Filtration Cap System
Various filtration-cap system accessories
Have Questions?
Catalog NumberType
FISK953914-0001Support Frit
Catalog number FISK953914-0001
Price (SGD)
229.00
EA
Add to cart
Type:
Support Frit
Price (SGD)
229.00
EA
Add to cart
  • Coarse-porosity glass support frit: 47mm (1.85 in.)
  • Upper screw clamp: 47mm (1.85 in.)
  • Drip tip: PTFE
  • Hose barb: PFA, 6.35mm (0.25 in.)
  • Filtration cap: GL 45
Specifications
For Use With (Equipment)Ultra-ware™ filtration cap system
No. per Case1 Case
TypeSupport Frit
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link