Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
Have Questions?
Catalog NumberVolume (Metric)
FISK953835-0000500 mL (Funnel), 2000 mL (Flask)
Catalog number FISK953835-0000
Price (SGD)
2,564.00
EA
Add to cart
Volume (Metric):
500 mL (Funnel), 2000 mL (Flask)
Price (SGD)
2,564.00
EA
Add to cart
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
  • The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
MaterialGlass
TypeMicrofiltration Assembly
Volume (Metric)500 mL (Funnel), 2000 mL (Flask)
Unit SizeEA