Kimble™ Kontes™ Filtration Assemblies: Borosilicate Glass Funnels for 47mm Assemblies
Kimble™ Kontes™ Filtration Assemblies: Borosilicate Glass Funnels for 47mm Assemblies
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Filtration Assemblies: Borosilicate Glass Funnels for 47mm Assemblies
Accessory for 47mm Assemblies
Have Questions?
Catalog NumberVolume (Metric)
FISK953771-0000500 mL
Catalog number FISK953771-0000
Price (SGD)
-
Volume (Metric):
500 mL
  • Accessory/ Replacement for: K953755-0000, K953805-0000, K953855-1047, K953825-0000, K953835-0000, K953845-0000
Specifications
For Use With (Equipment)47 mm Assemblies
MaterialBorosilicate Glass
Volume (Metric)500 mL
Unit SizeEA