Kimble™ KIMAX™ Heavy-Duty Clear Glass Class A Volumetric Flasks with Glass Stoppers
Kimble™ KIMAX™ Heavy-Duty Clear Glass Class A Volumetric Flasks with Glass Stoppers
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Heavy-Duty Clear Glass Class A Volumetric Flasks with Glass Stoppers
Large numbers make it easy to identify volume from far away
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISK92810W-5050 mL
FIS13-756-685200 mL
FIS13-756-6871000 mL
FIS13-756-6845 mL
FIS10-310-23620 mL
FIS13-756-6882000 mL
FIS10-212-7A100 mL
FIS10-310-23510 mL
FIS10-310-23725 mL
FIS13-756-686500 mL
Catalog number FISK92810W-50
Price (SGD)
137.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
137.00
PK
Add to cart
Large numbers make it easy to identify volume from far away
  • Wide Mouth
  • Heavy, uniform walls provide superior strength, durability and safety
  • Large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Calibrated “To Contain”
  • Capacity:10 to 250mL
  • Calibrated to meet ASTM™ E288
Specifications
Closure MaterialGlass
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Height (English)5.9 in.
Height (Metric)15 cm
Tolerance±0.08 mL
GraduatedYes
Mouth StyleWide
Closure Size13
Unit SizePK