1x1 image pixel for data collection
Tee connecting union for B/T 92 KYNAR (PVDF)
Product Image
DWK Life Sciences
Tee connecting union for B/T 92 KYNAR (PVDF)
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
Have Questions?
Catalog NumberDiameter (English) Inner
FISK749541-00001 in.
Catalog number FISK749541-0000
Price (SGD)
45.00
EA
Add to cart
Diameter (English) Inner:
1 in.
Price (SGD)
45.00
EA
Add to cart
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in a range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1 in. ID
Specifications
DescriptionBarbed fittings, T connector, Kynar, 1 in. ID, 25/32 in., 3-15/16 in., 2-19/32 in.;
Diameter (English) Inner1 in.
Diameter (English) Outer25/32 in.
Overall Length (English)4 in.
Packaging Quantity5/pk.
Outlet PortBarb
Sterilization MethodAutoclave or ethylene oxide (ETO)
Width (English)2 1/2 in.
Material (Seal)PVDF
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link