▸
Histology and Cytology
▸
Tissue Grinders
▸
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor
Produces more complete disaggregation than manual mixing.
Have Questions?
Catalog Number
Product Type
FISK749540-0000
Cordless Motor
Catalog number
FISK749540-0000
Price (SGD)
557.00
Each
Add to cart
Product Type:
Cordless Motor
Price (SGD)
557.00
Each
Add to cart
Produces more complete disaggregation than manual mixing
Use with either the disposable or reusable Pellet Pestle™ Mixers
Comes with two “AA” batteries
Specifications
For Use With (Equipment)
Pellet pestle mixer
Includes
2 AA batteries
Product Type
Cordless Motor