Check Order Status
Kimble™ Kontes™ Reusable Pellet Pestles™ Tissue Grinders
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Reusable Pellet Pestles™ Tissue Grinders
For use with 15mL micro centrifuge tubes
Have Questions?
Catalog Number
Capacity
FISK749515-1500
15 mL
Catalog number
FISK749515-1500
Price (SGD)
489.00
CS
Add to cart
Capacity:
15 mL
Price (SGD)
489.00
CS
Add to cart
For use with 15mL micro centrifuge tubes
Autoclavable CTFE/stainless-steel
Specially designed to match microtubes
Size: 15mL
Overall Pestle Length: 21cm
Specifications
Autoclavable
Autoclavable
Capacity
15 mL
For Use With (Equipment)
15mL Micro Tubes
Includes
Cs. of 1
Length (English)
8.26 in.
Length (Metric)
210mm
Material
CTFE/Stainless Steel