Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
Heavy wall design increases flask durability.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Tolerance
FISK623020-0400400 mL290 mm±0.15 mL
Catalog number FISK623020-0400
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
400 mL
Length (Metric):
290 mm
Tolerance:
±0.15 mL
Heavy wall design increases flask durability
  • Wide mouth mixing bulb provides easy access into the flask for dissolving tablets and other solids
  • Mixing bulb in neck accommodates approximately one tenth of the flask's total volume
Specifications
Capacity (Metric)400 mL
MaterialBorosilicate Glass
Height (English)11.41 in.
Height (Metric)29 cm
Length (Metric)290 mm
Neck StyleBulb, wide mouth
No. per CaseCase of 1
Tolerance±0.15 mL
Closure Size16
Unit SizeEA