Kimble™ Kontes™ Mojonnier™ Fat Extraction Flask
Kimble™ Kontes™ Mojonnier™ Fat Extraction Flask
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Mojonnier™ Fat Extraction Flask
For examination of dairy products
Have Questions?
Catalog NumberProduct Type
FISK617600-0025Fat Extraction Flask
Catalog number FISK617600-0025
Price (SGD)
345.00
EA
Add to cart
Product Type:
Fat Extraction Flask
Price (SGD)
345.00
EA
Add to cart
For examination of dairy products
  • Capacity: 25mL
Mojonnier Flask
  • Settling chamber at the lower portion has a capacity of ∽25mL
  • Diameter of extraction chamber is 1.375 in. (35mm)
  • Top opening has a pouring lip which is tooled for a rubber stopper
Mojonnier Threaded Flask
  • Supplied with a black, phenolic cap with PTFE-faced white rubber liner
  • GPI thread finish: 24-410
Specifications
DescriptionFlask. (Use with Size 0 stopper; sold separately.)
Capacity (Metric)25 mL
MaterialBorosilicate Glass
For Use With (Application)For fat extraction, examination of dairy products
Height (English)3.14 in.
Height (Metric)8 cm
Product TypeFat Extraction Flask
Closure TypeSize 0 stopper (sold separately)
Unit SizeEA