Check Order Status
Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
With or without fritted disc
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Column Capacity
Includes
FISK420300-0021
250 mm
22 mm
95 mL
Without fritted disc
Catalog number
FISK420300-0021
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric) Inner:
22 mm
Column Capacity:
95 mL
Includes:
Without fritted disc
Borosilicate glass
Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
For Use With (Application)
Many separations in adsorption and ion exchange chromatography
Diameter (Metric) Inner
22 mm
Type
Adsorption/Ion Exchange Chromatography Column
Column Capacity
95 mL
Length (Metric)
250 mm
Material
Borosilicate Glass
Support Material
Glass Wool
Includes
Without fritted disc