Kimble™ Kontes™ PTFE Class A Volumetric Flasks
Kimble™ Kontes™ PTFE Class A Volumetric Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ PTFE Class A Volumetric Flasks
Micro volumetric flask with an open top phenolic screw cap and a PTFE-lined silicone septum for syringe access.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISK297050-00022 mL
FISK297050-00011 mL
Catalog number FISK297050-0002
Price (SGD)
202.00
EA
Add to cart
Capacity (Metric):
2 mL
Price (SGD)
202.00
EA
Add to cart
  • Threaded flask
  • Open top phenolic cap
  • PTFE-lined silicone septum for syringe access
  • Calibrated “To Contain”
Specifications
CalibrationCalibrated To Contain
Capacity (English)0.067 oz.
Closure MaterialPhenolic
Capacity (Metric)2 mL
MaterialPTFE
Height (English)2.87 in.
Height (Metric)7.3 cm
IncludesPTFE-lined Silicone septum
Neck StyleThreaded
Tolerance±0.015 mL
Bottom ShapeV-shape
Closure Size13-425
Unit SizeEA