Kimble™ Kontes™ Distillation Receiver Graduated Tube
Kimble™ Kontes™ Distillation Receiver Graduated Tube
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Distillation Receiver Graduated Tube
Useful as a centrifuge or extraction tube as well as a receiver.
Have Questions?
Catalog number FISK288251-0000
Price (SGD)
-
  • Distillation Receiver Graduated Tube
  • 14/20 Standard Taper Joint Size
  • Capacity 12mL
  • Grad. x Subdiv. 0-2 x 0.1mL and 2-12 x 0.2mL
  • Overall Length 142mm
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Uses Standard Taper polyacetal clamps, not included
Specifications
Product TypeDistillation Receiver Tube
Capacity (Metric)12 mL
Capacity (English)0.4 oz.
Graduations0 to 2 mL/0.1 mL, 2 to 12 mL/0.2 mL
IncludesWithout hooks for clamp attachment
For Use With (Application)Useful as a centrifuge or extraction tube as well as a receiver
Standard Taper Joint14/20
Length (English)5.59 in.
Length (Metric)14.2 cm
Unit SizeEA