Immobilon™-FL PVDF Membranes
Immobilon™-FL PVDF Membranes
Actual product may vary
MilliporeSigma
Immobilon™-FL PVDF Membranes
Designed specifically for Western blots and protein sequencing. Immobilon™-FL PVDF Transfer Membranes are plain surfaced with 0.45 μm pores and have lower background fluorescence than other membranes. Available in 10 and 20 cm squares and a 3.75 m roll.
Have Questions?
Catalog NumberFormatDimensions (LxW)Quantity
FISIPFL10100Sheet10 x 10 cm (3.9 x 3.9 in.)10 Sheets
Catalog number FISIPFL10100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Format:
Sheet
Dimensions (LxW):
10 x 10 cm (3.9 x 3.9 in.)
Quantity:
10 Sheets
  • The first transfer membrane optimized for fluorescence applications
  • Extremely low background improves sensitivity of all fluorescence detection protocols
  • Compatible with all commonly used fluorescent probes at all excitation and emission wavelengths
  • Ideal for multiplexing and chemifluorescence applications
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications
Length (Metric)10 cm
Width (Metric)10 cm
For Use With (Application)Fluorescence, Chemifluorescence, Chromogenic, Chemiluminescence
ColorWhite
CodeIPFL
Quantity10 Sheets
Width (English)3.9 in.
FormatSheet
MaterialPVDF
Length (English)3.9 in.
Pore Size0.45 μm
Dimensions (LxW)10 x 10 cm (3.9 x 3.9 in.)
Unit SizePK