Express™ PLUS Membrane Filters
Express™ PLUS Membrane Filters
Actual product may vary
MilliporeSigma
Express™ PLUS Membrane Filters
Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.
Have Questions?
Catalog NumberPore SizeDiameter (Metric)
FISGPWP047000.22 μm47 mm
Catalog number FISGPWP04700
Price (SGD)
717.00
PK
Add to cart
Pore Size:
0.22 μm
Diameter (Metric):
47 mm
Price (SGD)
717.00
PK
Add to cart
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
Features and Benefit:
  • Ultra fast filtration and high throughput shorten process time
  • Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
  • Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specifications
AutoclavableNot Autoclavable
ColorWhite
DisposableReusable
Filter TypeScreen Filter
FormatDisc
MaterialPES
Pore Size0.22 μm
SterilityNon-sterile
Flow Rate15 (at 690 Mbar (10 psi))
For Use With (Application)Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
HydrophobicityHydrophilic
Membrane TypePlain
TypeFilter Membranes
WettabilityHydrophilic
Diameter (English)1.9 in.
Diameter (Metric)47 mm
Unit SizePK