Porcelain Buchner Funnels with Fixed Perforated Plates
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) DiscDiameter (Metric) Plate
FISFB966M4500 mL240mm280mm
FISFB966A2 mL13mm14.5mm
FISFB966B30 mL35 - 40mm41 mm
FISFB966C87 mL50 - 55mm56mm
FISFB966D186 mL70mm83mm
FISFB966F320 mL90mm100mm
FISFB966G550 mL110mm114mm
FISFB966H700 mL110 - 124mm126mm
FISFB966J1060 mL142 - 150mm151mm
FISFB966K1860 mL185mm186mm
Catalog number FISFB966M
Price (SGD)
763.00
CS
Add to cart
Capacity (Metric):
4500 mL
Diameter (Metric) Disc:
240mm
Diameter (Metric) Plate:
280mm
Price (SGD)
763.00
CS
Add to cart
For suction filtration applications
  • Autoclavable
  • Highly resistant to chemical attack
  • Without cover
  • Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top280mm
Capacity (Metric)4500 mL
MaterialPorcelain
Height (Metric)35 cm
AutoclavableAutoclavable
Diameter (Metric) Plate280mm
For Use With (Application)Suction filtration applications
Diameter (Metric) Disc240mm
IncludesFixed perforated plate
QuantityEach
Wall Thickness6.8mm
Weight (Metric)4.2kg
Product TypeBuchner Funnel
Unit SizeCS