Wide-Form Porcelain Crucibles
Wide-Form Porcelain Crucibles
Actual product may vary
Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)Weight (Metric)
FISFB965P250 mL102mm60mm124g
FISFB965I8 mL32mm20mm10g
FISFB965K30 mL50mm31mm26g
FISFB965M100 mL76mm46mm55g
Catalog number FISFB965P
Price (SGD)
202.00
PK
Add to cart
Capacity (Metric):
250 mL
Diameter (Metric) Outer:
102mm
Height (Metric):
60mm
Weight (Metric):
124g
Price (SGD)
202.00
PK
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2.3mm
Weight (Metric)124g
Capacity (Metric)250 mL
Diameter (Metric) Outer102mm
Height (Metric)60mm
Product TypeCrucible
Unit SizePK