Porcelain Mortars
Porcelain Mortars
Actual product may vary
Porcelain Mortars
Glazed outside except bottom and unglazed inside
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityDiameter (English) OuterDiameter (Metric) Outer
FISFB961H37.19 oz., 1100mL7.32 in.18.6cm
FISFB961A1.69 oz., 50mL2.75 in.7cm
FISFB961D9.29 oz., 275mL4.52 in.11.5cm
FISFB961E13.5 oz., 400mL5.11 in.13cm
FISFB961F25.3 oz., 750mL6.29 in.16cm
Catalog number FISFB961H
Price (SGD)
336.00
EA
Add to cart
Capacity:
37.19 oz., 1100mL
Diameter (English) Outer:
7.32 in.
Diameter (Metric) Outer:
18.6cm
Price (SGD)
336.00
EA
Add to cart
Glazed outside except bottom and unglazed inside
  • With lip
  • Deep form
  • Export quality
  • Color: white
  • Available in various capacities
Specifications
Capacity (Metric)1100 mL
FinishGlazed Outer
Capacity (English)37.19 oz.
Diameter (English) Outer7.32 in.
Diameter (Metric) Outer18.6cm
Product TypeMortar
MaterialPorcelain
Capacity37.19 oz., 1100mL
Unit SizeEA