Alumina Crucibles
Alumina Crucibles
Actual product may vary
Alumina Crucibles
Made up of 99.7% Alumina
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Product Type
FISFB960K50 mLHigh-Form Crucible
FISFB960A2 mLCylindrical form crucible
FISFB960I10 mLHigh-Form Crucible
Catalog number FISFB960K
Price (SGD)
170.00
EA
Add to cart
Capacity (Metric):
50 mL
Product Type:
High-Form Crucible
Price (SGD)
170.00
EA
Add to cart
Made up of 99.7% Alumina
  • Available in two forms: high and cyclindrical
  • Unglazed
  • Maximum use temperature: 1750°C
  • Resistant to Alkalies, chemical attack and other fluxes
  • Inert in hydrogen and carbonaceous atmospheres
  • Color: cream
Specifications
ColorCream
MaterialAlumina
Max. Temperature (Metric)1750°C
Capacity (Metric)50 mL
Diameter (Metric) Outer48mm
Height (Metric)47mm
Product TypeHigh-Form Crucible
ShapeCylinder
Unit SizeEA