Porcelain Bitumen Filtering Crucible
Porcelain Bitumen Filtering Crucible
Actual product may vary
Porcelain Bitumen Filtering Crucible
Filtration device for general laboratory use
Have Questions?
Catalog number FISFB960G
Price (SGD)
53.00
EA
Add to cart
Price (SGD)
53.00
EA
Add to cart
  • Porcelain made
  • Color: white
  • Capacity: 28mL
Specifications
ColorWhite
Diameter (Metric) Inner31.3mm
MaterialPorcelain
For Use With (Application)General laboratory use
Thickness (Metric)1.7mm
Capacity (Metric)28 mL
Diameter (Metric) Outer33mm
Height (Metric)29mm
Product TypeBitumen Filtering Crucible
Unit SizeEA