High-Form Alumina Crucibles Covers
Actual product may vary
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
Covers for High-Form Alumina crucibles
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
FISFB9601For 10mL crucible
FISFB9603For 50mL crucible
Catalog number FISFB9601
Price (SGD)
-
For Use With (Application):
For 10mL crucible
  • For 0.3, 0.6, 1.6 and 3.3 oz. (10, 20, 50 and 100mL) crucibles
  • Alumina construction
  • Color: cream
Specifications
ColorCream
MaterialAlumina
For Use With (Application)For 10mL crucible
Product TypeCrucible Cover
Unit SizePack