Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FISFB800250 | 250 mL |
FISFB800100 | 100 mL |
FKS15446113 | 100 mL |
FISFB-800-250 | 250 mL |
FKS15456113 | 250 mL |
FISFB-800-500 | 500 mL |
FISFB800500 | 500 mL |
FKIFB800-500 | 500 mL |
FKS15476113 | 500 mL |
FISFB-800-1000 | 1000 mL |
FISFB8001000 | 1000 mL |
FKS15486113 | 1000 mL |
FISFB8002000 | 2000 mL |
FKS15406123 | 2000 mL |
FISFB8005000 | 5000 mL |
FKS15416123 | 5000 mL |
FISFB80010000 | 10,000 mL |
FKS15436123 | 10,000 mL |