Check Order Status
Reusable Glass Rim Top Culture Tubes
Actual product may vary
Reusable Glass Rim Top Culture Tubes
Uniform wall thickness for maximum heat transfer and chemical resistance
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Length (Metric)
Diameter (Metric)
FISFB70016125
20 mL
125 mm
16 mm
FISFB70013100
10 mL
100 mm
13 mm
2 Options
Catalog number
FISFB70016125
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
20 mL
Length (Metric):
125 mm
Diameter (Metric):
16 mm
Made from KG-33 borosilicate glass Durable, white ceramic enamel marking spot
Fire-polished top rim
Specifications
Certifications/Compliance
USP Type I, ASTM E 438 Type I, Class A
Color
Clear
Capacity (Metric)
20 mL
Length (Metric)
125 mm
Material
Borosilicate Glass
Label Type
White Ceramic Enamel Marking Spot
Sterility
Non-sterile
Autoclavable
Autoclavable
Diameter (Metric)
16 mm
Closure Type
Not Included
Product Type
Culture Tube