Reusable Glass Long-Stem Funnels
Reusable Glass Long-Stem Funnels
Actual product may vary
Reusable Glass Long-Stem Funnels
Promote faster, more effective filtering
Have Questions?
Catalog NumberCapacity (Metric)
FISFB60058100185 mL
Catalog number FISFB60058100
Price (SGD)
208.00
PK
Add to cart
Capacity (Metric):
185 mL
Price (SGD)
208.00
PK
Add to cart
  • Constructed with heavy uniform walls, molded rims and fire-polished stems
  • Formed to a 58° angle for rapid filtration
  • Cone is suspended by its uppermost edge, leaving most of the conical area for filtration
  • High resistance to chemical attack, mechanical and thermal shock
Specifications
Diameter (Metric)Top10cm
Capacity (Metric)185 mL
Diameter (Metric) Stem9.57 in.
MaterialBorosilicate Glass
Height (Metric)243 mm
Certifications/ComplianceASTM E1095, Type I, Class B
Angle58°
Diameter (Metric) Plate100mm
Diameter (Metric) Filter18.5cm
For Use With (Application)Provides faster, more effective filtering
Length (Metric) Stem15cm
Unit SizePK