Reusable Glass Wide-Mouth Erlenmeyer Flasks
Actual product may vary
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduation RangeHeight (English)Closure Size
FISFB501125125 mL50 to 125 mL4.25 in.6
FISFB50120002000 mL600 to 2000 mL10.4 in.13
FISFB501500500 mL100 to 500 mL6.77 in.10
FISFB501250250 mL50 to 225 mL5.11 in.8
Catalog number FISFB501125
Price (SGD)
173.00
Pack
Capacity (Metric):
125 mL
Graduation Range:
50 to 125 mL
Height (English):
4.25 in.
Closure Size:
6
Price (SGD)
173.00
Pack
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
  • Capacity scale measures approximate volume
  • Matte-finish ceramic marking spot takes writing and erases easily
Specifications
DescriptionTop style: Heavy duty tooled with capacity scale
Diameter (English) Outer2.59 in.
Capacity (Metric)125 mL
MaterialBorosilicate Glass
Graduation Range50 to 125 mL
Graduations25 mL
Height (English)4.25 in.
Height (Metric)10.8 cm
No. of Positions1
Writing SurfaceMatte Finish Ceramic Enamel Marking
DisposableReusable
Mouth StyleWide
Diameter (Metric) Outer66 mm
Closure Size6
Unit SizePack