Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Height (Metric) | Graduation Range | Closure Size |
|---|---|---|---|---|
| FISFB300500 | 500 mL | 19 cm | 150 to 500 mL | 7 |
| FISFB3002000 | 2000 mL | 30 cm | 600 to 2000 mL | 9 |
| FISFB3001000 | 1000 mL | 23.8 cm | 300 to 1000 mL | 8 |
| FISFB300250 | 250 mL | 16.3 cm | 50 to 250 mL | 6 |