Reusable Heavy-Wall Filter Flasks
Actual product may vary
Reusable Heavy-Wall Filter Flasks
With heavier walls than standard Erlenmeyer flasks
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Graduation RangeClosure Size
FISFB300250250 mL16.3 cm50 to 250 mL6
FISFB30020002000 mL30 cm600 to 2000 mL9
FISFB30010001000 mL23.8 cm300 to 1000 mL8
FISFB300500500 mL19 cm150 to 500 mL7
Catalog number FISFB300250
Price (SGD)
265.00
Pack
Capacity (Metric):
250 mL
Height (Metric):
16.3 cm
Graduation Range:
50 to 250 mL
Closure Size:
6
Price (SGD)
265.00
Pack
With heavier walls than standard Erlenmeyer flasks
  • Side tubulation Capacity scale
  • Designed to take 1 atmosphere of vacuum or 29 in. of mercury
  • Sizes 25 through 500mL have side hose connection designed to accept 5.16 in. O.D. flexible tubing
  • Sizes 1000mL and larger accept 3.8 in. O.D. tubing
  • Temperature range: 840°F (450°C) max.
Specifications
Certifications/ComplianceASTM E 1406, Type II, Class I
Diameter (English) Outer3.26 in.
Diameter (Metric) Inner Tubing0.78 cm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Graduation Range50 to 250 mL
Graduations25 mL
Height (English)6.4 in.
Height (Metric)16.3 cm
Temperature Range (English)840°F max.
GraduatedYes
ClassClass I
Diameter (Metric) Outer8.3 cm
Closure Size6
Unit SizePack