Round-Bottom Polypropylene Test Tubes With Cap
Actual product may vary
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityGraduatedPackaging
FISFB149561D5 mLSterileNoBlister Pk.
FISFB149569C14 mLSterileYesBlister Pack
Catalog number FISFB149561D
Price (SGD)
121.00
Case
Capacity (Metric):
5 mL
Sterility:
Sterile
Graduated:
No
Packaging:
Blister Pk.
Price (SGD)
121.00
Case
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without a snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
ColorTranslucent
Capacity (Metric)5 mL
Length (Metric)75 mm
MaterialPolypropylene
DisposableSingle-use
PackagingBlister Pk.
SterilitySterile
Product LineFisherbrand
GraduatedNo
Closure TypeSnap Cap
Product TypeTest Tube
Unit SizeCase