Pipet Controller
Pipet Controller
Actual product may vary
Pipet Controller
Pipet Controller
Pipet Controller
A powerful, lightweight and ergonomically designed pipetting controller for all commonly used glass or plastic pipettes
Have Questions?
Catalog number FISFB14955202
Price (SGD)
1,036.00
EA
Add to cart
Price (SGD)
1,036.00
EA
Add to cart
  • Powerful yet quiet pump
  • Ergonomic design
  • Soft grip push buttons
  • Double safety valve
  • Autoclavable pipet cone
  • 3-speed modes High, Low and Gravity
  • Switch located on slide part of controller ensuring ease of operation
  • LED indicator lights up in red when battery is low
  • Can be operated continuously for 8 hours
  • Replaceable membrane Filter to protect device from contaminants being drawn into pipet
  • Can be used while it is charging
Specifications
Battery Type1000mAh NiMH batteries
Compatible TipsCone shape
ColorBlue
Grip TypeSoft Push Button
TypePipette Controller
Volume (Metric)0.1 to 100 mL
Filter TypeHydrophobic PTFE membrane filter
Finger StylePush button
For Use With (Equipment)Glass or plastic from 0.1mL to 100mL
IncludesPipet Filler, Charger, Desktop Stand, Wall mount bracket, Two spare membrane filters: 0.20μm and 0.45μm, Instruction manual, Warranty Card, batteries
Number of Speeds3
Recharge Time2 to 2 hours
Electrical Requirements2 x 1.2V/ 1000mAh NiMH batteries
Voltage2 x 1.2 V
Unit SizeEA